Anti-CLEC5A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 226402]

Supplier: R & D Systems
FAB1639P
220017-152EA 513.38 USD
220017-152
Anti-CLEC5A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 226402]
Antibodies
Type: Primary
Antigen: CLEC5A
Clonality: Monoclonal
Clone: 226402
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR