Anti-CLEC4M Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 120604]

Supplier: R & D Systems
FAB162P
220017-149EA 485.91 USD
220017-149
Anti-CLEC4M Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 120604]
Antibodies
Type: Primary
Antigen: CLEC4M
Clonality: Monoclonal
Clone: 120604
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR