Anti-CD209 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 120612]

Supplier: R & D Systems
FAB1621P
220017-147EA 513.38 USD
220017-147
Anti-CD209 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 120612]
Antibodies
Type: Primary
Antigen: CD209
Clonality: Monoclonal
Clone: 120612
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR