Anti-CXCR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 49801]

Supplier: R & D Systems
FAB160P-025 FAB160P-100
102881-258EA 205.35 USD
102881-258 102881-260
Anti-CXCR3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 49801]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 49801
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR