Anti-CXCR3 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 49801]

Supplier: R & D Systems
FAB160N-100 FAB160N-025
102881-256EA 519.17 USD
102881-256 102880-726
Anti-CXCR3 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 49801]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 49801
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR