Anti-CXCR3 Mouse Monoclonal Antibody (Fluorescein) [clone: 49801]

Supplier: R & D Systems
FAB160F-100 FAB160F-025
102880-720EA 177.88 USD
102880-720 102880-722
Anti-CXCR3 Mouse Monoclonal Antibody (Fluorescein) [clone: 49801]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 49801
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR