Anti-CXCR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 49801]

Supplier: R & D Systems
FAB160A-100 FAB160A-025
102880-712EA 202.46 USD
102880-712 102880-714
Anti-CXCR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 49801]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 49801
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR