Anti-MICB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 236511]

Supplier: R & D Systems
FAB1599P-025 FAB1599P-100
102880-702EA 229.94 USD
102880-702 102880-704
Anti-MICB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 236511]
Antibodies
Type: Primary
Antigen: MICB
Clonality: Monoclonal
Clone: 236511
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR