Anti-MICB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 236511]

Supplier: R & D Systems
FAB1599A-100 FAB1599A-025
102880-264EA 582.8 USD
102880-264 102880-262
Anti-MICB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 236511]
Antibodies
Type: Primary
Antigen: MICB
Clonality: Monoclonal
Clone: 236511
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR