Anti-CCR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 205410]

Supplier: R & D Systems
FAB1567P-100 FAB1567P-025
102880-258EA 205.35 USD
102880-258 102880-260
Anti-CCR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 205410]
Antibodies
Type: Primary
Antigen: CCR4
Clonality: Monoclonal
Clone: 205410
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR