Anti-CCR4 Mouse Monoclonal Antibody (Fluorescein) [clone: 205410]

Supplier: R & D Systems
FAB1567F-100 FAB1567F-025
102880-254EA 202.46 USD
102880-254 102880-256
Anti-CCR4 Mouse Monoclonal Antibody (Fluorescein) [clone: 205410]
Antibodies
Type: Primary
Antigen: CCR4
Clonality: Monoclonal
Clone: 205410
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR