Anti-CCR4 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 205410]

Supplier: R & D Systems
FAB1567C-025 FAB1567C-100
102880-250EA 219.82 USD
102880-250 102880-252
Anti-CCR4 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 205410]
Antibodies
Type: Primary
Antigen: CCR4
Clonality: Monoclonal
Clone: 205410
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR