Anti-CCR4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 205410]

Supplier: R & D Systems
FAB1567A-025 FAB1567A-100
102880-246EA 219.82 USD
102880-246 102880-248
Anti-CCR4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 205410]
Antibodies
Type: Primary
Antigen: CCR4
Clonality: Monoclonal
Clone: 205410
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR