Anti-OLR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 214012]

Supplier: R & D Systems
FAB1564P
220017-128EA 533.63 USD
220017-128
Anti-OLR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 214012]
Antibodies
Type: Primary
Antigen: OLR1
Clonality: Monoclonal
Clone: 214012
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR