Anti-CCR3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 61828]

Supplier: R & D Systems
FAB155P-100 FAB155P-025
102880-232EA 193.78 USD
102880-232 102880-234
Anti-CCR3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 61828]
Antibodies
Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: 61828
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR