Anti-CCR3 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 61828]

Supplier: R & D Systems
FAB155C-100 FAB155C-025
102880-630EA 561.11 USD
102880-630 102880-628
Anti-CCR3 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 61828]
Antibodies
Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: 61828
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR