Anti-CCR3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 61828]

Supplier: R & D Systems
FAB155A-025 FAB155A-100
102880-624EA 202.46 USD
102880-624 102880-626
Anti-CCR3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 61828]
Antibodies
Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: 61828
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR