Anti-NOTCH3 Sheep Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB1559A
102881-254EA 552.43 USD
102881-254
Anti-NOTCH3 Sheep Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: NOTCH3
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR