Anti-PODXL Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 192703]

Supplier: R & D Systems
FAB1556P
220017-123EA 513.38 USD
220017-123
Anti-PODXL Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 192703]
Antibodies
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 192703
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR