Anti-PODXL Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 192703]

Supplier: R & D Systems
FAB1556A
220017-122EA 577.02 USD
220017-122
Anti-PODXL Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 192703]
Antibodies
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 192703
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR