Anti-AXL Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 108724]

Supplier: R & D Systems
FAB154P
102881-250EA 582.8 USD
102881-250
Anti-AXL Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 108724]
Antibodies
Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 108724
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR