Anti-AXL Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 108724]

Supplier: R & D Systems
FAB154C
102880-556EA 633.42 USD
102880-556
Anti-AXL Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 108724]
Antibodies
Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 108724
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR