Anti-AXL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 108724]

Supplier: R & D Systems
FAB154A
102880-554EA 639.2 USD
102880-554
Anti-AXL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 108724]
Antibodies
Type: Primary
Antigen: AXL
Clonality: Monoclonal
Clone: 108724
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR