Anti-INSR Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB1544P
102880-540EA 501.82 USD
102880-540
Anti-INSR Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: INSR
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR