Anti-INSR Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB1544G
102880-536EA 501.82 USD
102880-536
Anti-INSR Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: INSR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR