Anti-INSR Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB1544A
220017-118EA 543.75 USD
220017-118
Anti-INSR Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: INSR
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR