Anti-TDGF1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 237603]

Supplier: R & D Systems
FAB1538A
102880-534EA 577.02 USD
102880-534
Anti-TDGF1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 237603]
Antibodies
Type: Primary
Antigen: TDGF1
Clonality: Monoclonal
Clone: 237603
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR