Anti-TLR6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 418601]

Supplier: R & D Systems
FAB1533P
220017-116EA 513.38 USD
220017-116
Anti-TLR6 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 418601]
Antibodies
Type: Primary
Antigen: TLR6
Clonality: Monoclonal
Clone: 418601
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR