Anti-TLR6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 418601]

Supplier: R & D Systems
FAB1533A
220017-115EA 561.11 USD
220017-115
Anti-TLR6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 418601]
Antibodies
Type: Primary
Antigen: TLR6
Clonality: Monoclonal
Clone: 418601
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR