Anti-TLR2 Rat Monoclonal Antibody (Fluorescein) [clone: 203325]

Supplier: R & D Systems
FAB1530F
220017-114EA 420.83 USD
220017-114
Anti-TLR2 Rat Monoclonal Antibody (Fluorescein) [clone: 203325]
Antibodies
Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: 203325
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR