Anti-HAVCR2 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 215008]

Supplier: R & D Systems
FAB1529G
102880-532EA 600.15 USD
102880-532
Anti-HAVCR2 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 215008]
Antibodies
Type: Primary
Antigen Symbol: HAVCR2
Antigen Name: hepatitis A virus cellular receptor 2
Antigen Synonyms: TIM-3, Tim3, Timd3
Clonality: Monoclonal
Clone: 215008
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR