Anti-CLEC4N Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB1525A
220017-110EA 577.02 USD
220017-110
Anti-CLEC4N Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: CLEC4N
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR