Anti-CCR2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 48607]

Supplier: R & D Systems
FAB151N-025 FAB151N-100
102880-522EA 219.82 USD
102880-522 102880-524
Anti-CCR2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 48607]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 48607
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR