Anti-CCR2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 48607]

Supplier: R & D Systems
FAB151G-100 FAB151G-025
102880-520EA 594.37 USD
102880-520 102880-518
Anti-CCR2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 48607]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 48607
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR