Anti-CCR2 Mouse Monoclonal Antibody (Biotin) [clone: 48607]

Supplier: R & D Systems
FAB151B
220017-107EA 480.12 USD
220017-107
Anti-CCR2 Mouse Monoclonal Antibody (Biotin) [clone: 48607]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 48607
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR