Anti-CCR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 48607]

Supplier: R & D Systems
FAB151A-100 FAB151A-025
102880-510EA 227.05 USD
102880-510 102880-512
Anti-CCR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 48607]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 48607
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR