Anti-ULBP3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 166510]

Supplier: R & D Systems
FAB1517P
102880-508EA 600.15 USD
102880-508
Anti-ULBP3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 166510]
Antibodies
Type: Primary
Antigen: ULBP3
Clonality: Monoclonal
Clone: 166510
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR