Anti-DLL4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 447506]

Supplier: R & D Systems
FAB1506P
102880-502EA 513.38 USD
102880-502
Anti-DLL4 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 447506]
Antibodies
Type: Primary
Antigen: DLL4
Clonality: Monoclonal
Clone: 447506
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR