Anti-LRP6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 255302]

Supplier: R & D Systems
FAB1505A
102880-498EA 608.83 USD
102880-498
Anti-LRP6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 255302]
Antibodies
Type: Primary
Antigen: LRP6
Clonality: Monoclonal
Clone: 255302
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR