Anti-IL27RA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 191106]

Supplier: R & D Systems
FAB14791G-100 FAB14791G-025
102880-488EA 561.11 USD
102880-488 102880-486
Anti-IL27RA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 191106]
Antibodies
Type: Primary
Antigen: IL27RA
Clonality: Monoclonal
Clone: 191106
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR