Anti-IL13RA1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 419718]

Supplier: R & D Systems
FAB1462N
102880-474EA 543.75 USD
102880-474
Anti-IL13RA1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 419718]
Antibodies
Type: Primary
Antigen: IL13RA1
Clonality: Monoclonal
Clone: 419718
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR