Anti-CCR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53504]

Supplier: R & D Systems
FAB145P-100 FAB145P-025
102881-244EA 219.82 USD
102881-244 102881-246
Anti-CCR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53504]
Antibodies
Type: Primary
Antigen: CCR1
Clonality: Monoclonal
Clone: 53504
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR