Anti-PTPRC Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 2D1]

Supplier: R & D Systems
FAB1430G
102881-220EA 342.74 USD
102881-220
Anti-PTPRC Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 2D1]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 2D1
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR