Anti-CCR8 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 191704]

Supplier: R & D Systems
FAB1429P-100 FAB1429P-025
102881-214EA 205.35 USD
102881-214 102881-216
Anti-CCR8 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 191704]
Antibodies
Type: Primary
Antigen: CCR8
Clonality: Monoclonal
Clone: 191704
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR