Anti-CCR8 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 191704]

Supplier: R & D Systems
FAB1429C-100 FAB1429C-025
102881-210EA 219.82 USD
102881-210 102881-212
Anti-CCR8 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 191704]
Antibodies
Type: Primary
Antigen: CCR8
Clonality: Monoclonal
Clone: 191704
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR