Anti-SLC2A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 202915]

Supplier: R & D Systems
FAB1418P
220017-072EA 549.54 USD
220017-072
Anti-SLC2A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 202915]
Antibodies
Type: Primary
Antigen: SLC2A1
Clonality: Monoclonal
Clone: 202915
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR