Anti-SLC2A2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 199017]

Supplier: R & D Systems
FAB1414P
102881-196EA 543.75 USD
102881-196
Anti-SLC2A2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 199017]
Antibodies
Type: Primary
Antigen: SLC2A2
Clonality: Monoclonal
Clone: 199017
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR