Anti-IL23R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 218213]

Supplier: R & D Systems
FAB14001P-025 FAB14001P-100
102881-192EA 310.92 USD
102881-192 102881-194
Anti-IL23R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 218213]
Antibodies
Type: Primary
Antigen: IL23R
Clonality: Monoclonal
Clone: 218213
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR