Anti-KLRK1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 149810]

Supplier: R & D Systems
FAB139P-025 FAB139P-100
102881-174EA 549.54 USD
102881-174 102881-172
Anti-KLRK1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 149810]
Antibodies
Type: Primary
Antigen: KLRK1
Clonality: Monoclonal
Clone: 149810
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR