Anti-KLRK1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 149810]

Supplier: R & D Systems
FAB139N-100 FAB139N-025
102881-168EA 177.88 USD
102881-168 102881-170
Anti-KLRK1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 149810]
Antibodies
Type: Primary
Antigen: KLRK1
Clonality: Monoclonal
Clone: 149810
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR